missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Isoliquiritigenin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001812
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001812 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001812 Supplier Merck Emd Millipore Supplier No. PHL8954410MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 961-29-5
Product Line phyproof
Linear Formula C15H12O4
MDL Number MFCD00075907
Synonym (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one; 4,2′,4′-Trihydroxychalcone
Type Reference Substance
Quantity 10 mg
Formula Weight 256.25
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG