accessibility menu, dialog, popup

UserName

CAS RN 96-49-1

IUPAC Name: 1,3-dioxolan-2-one
Synonyms: ethylene carbonate
Molecular Weight (g/mol): 88.06
Molecular Formula: C3H4O3
InChi Key: KMTRUDSVKNLOMY-UHFFFAOYSA-N
SMILES: O=C1OCCO1
Boiling Point 
  • (1)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)