accessibility menu, dialog, popup

UserName

OHClCl

CAS RN 95-77-2

IUPAC Name: 3,4-dichlorophenol
Synonyms: 3,4-dichlorophenol C6H4cl2O
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: WDNBURPWRNALGP-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)