CAS RN 95-26-1
IUPAC Name:
2,5-dimethyl-1,3-benzothiazole
Synonyms:
benzothiazole, 2,5-dimethyl-
Molecular Weight (g/mol):
163.24
Molecular Formula:
C9H9NS
InChi Key:
XHANCLXYCNTZMM-UHFFFAOYSA-N
SMILES:
CC1=NC2=CC(C)=CC=C2S1
1
–
1
of
1
results
1
2,5-Dimethylbenzothiazole 98.0+%, TCI America™
CAS: 95-26-1 Molecular Formula: C9H9NS Molecular Weight (g/mol): 163.238 MDL Number: MFCD00005796 InChI Key: XHANCLXYCNTZMM-UHFFFAOYSA-N Synonym: 2,5-dimethylbenzothiazole,benzothiazole, 2,5-dimethyl,2,5-dimethylbenzthiazol,2,5-dimethylbenzthiazol czech,benzothiazole, 2,5-dimethyl-6ci,7ci,8ci,9ci,acmc-209rya,2,5-dimethyl-benzothiazol,2,5-dimethyl benzothiazole,benzothiazole,2,5-dimethyl,4-27-00-01101 beilstein handbook reference PubChem CID: 7227 IUPAC Name: 2,5-dimethyl-1,3-benzothiazole SMILES: CC1=CC2=C(C=C1)SC(=N2)C