missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzothiazole 98.0+%, TCI America™

Catalog No. D121810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D121810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D121810G Supplier TCI America Supplier No. D121810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-26-1
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00005796
InChI Key XHANCLXYCNTZMM-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzothiazole, benzothiazole, 2,5-dimethyl, 2,5-dimethylbenzthiazol, 2,5-dimethylbenzthiazol czech, benzothiazole, 2,5-dimethyl-6ci,7ci,8ci,9ci, acmc-209rya, 2,5-dimethyl-benzothiazol, 2,5-dimethyl benzothiazole, benzothiazole,2,5-dimethyl, 4-27-00-01101 beilstein handbook reference
PubChem CID 7227
IUPAC Name 2,5-dimethyl-1,3-benzothiazole
SMILES CC1=CC2=C(C=C1)SC(=N2)C

Specifications

Melting Point 42°C
Boiling Point 146°C
Quantity 10 g
Formula Weight 163.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethylbenzothiazole
EINECSNumber (5)-0229
RTECSNumber DL3675000
TSCA Yes