accessibility menu, dialog, popup

UserName

CAS RN 942-91-6

HOOSS

CAS RN 942-91-6

IUPAC Name: 2-(benzenecarbothioylsulfanyl)acetic acid
Synonyms: (benzenecarbothioylsulfanyl)acetic acid
Molecular Weight (g/mol): 212.28
Molecular Formula: C9H8O2S2
InChi Key: XBEIANFIOZTEDE-UHFFFAOYSA-N
SMILES: OC(=O)CSC(=S)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results