accessibility menu, dialog, popup

UserName

CAS RN 94159-32-7

H3COOSNH3C

CAS RN 94159-32-7

IUPAC Name: 2-(4-methyl-1,3-thiazol-5-yl)ethyl hexanoate
Synonyms: 2-(4-methyl-1,3-thiazol-5-yl)ethyl hexanoate
Molecular Weight (g/mol): 241.35
Molecular Formula: C12H19NO2S
InChi Key: VJULDCZELAIZHC-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCCC1=C(C)N=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results