missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methyl-5-thiazolyl)ethyl Hexanoate 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 152°C |
| Quantity | 5 g |
| Formula Weight | 241.35 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(4-Methyl-5-thiazolyl)ethyl Hexanoate |
Safety and Handling
| TSCA | No |