accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 94-30-4

IUPAC Name: ethyl 4-methoxybenzoate
Synonyms: ethyl anisate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: FHUODBDRWMIBQP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results