accessibility menu, dialog, popup

UserName

OHSBr

CAS RN 937795-99-8

IUPAC Name: [4-(3-bromothiophen-2-yl)phenyl]methanol
Synonyms: [4-(3-bromothiophen-2-yl)phenyl]methanol
Molecular Weight (g/mol): 269.16
Molecular Formula: C11H9BrOS
InChi Key: AQBJKGXCOBEPMC-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)