missing translation for 'onlineSavingsMsg'
Learn More

[4-(3-Bromothien-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. p-3586561
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67809DA 1g Amber Glass Bottle
CC67809DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67809DA Supplier Thermo Scientific Chemicals Supplier No. CC67809DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 937795-99-8
Molecular Formula C11H9BrOS
Molecular Weight (g/mol) 269.156
MDL Number MFCD09879972
InChI Key AQBJKGXCOBEPMC-UHFFFAOYSA-N
Synonym 4-3-bromothien-2-yl phenyl methanol, 4-3-bromothiophen-2-yl phenyl methanol, 4-3-bromo-2-thienyl phenyl methanol
PubChem CID 24229755
IUPAC Name [4-(3-bromothiophen-2-yl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C2=C(C=CS2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 269.16
Percent Purity 97%
Chemical Name or Material [4-(3-Bromothien-2-yl)phenyl]methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.