accessibility menu, dialog, popup

UserName

CH3ONOOOH3COOCH3OOCH3OOH3CSOO

CAS RN 934591-76-1

IUPAC Name: methyl 3-acetyl-2-oxo-6-(phenylsulfanyl)-4-[1,2,3-tris(acetyloxy)propyl]-hexahydro-2H-pyrano[3,4-d][1,3]oxazole-6-carboxylate
Synonyms: methyl 3-acetyl-2-oxo-6-(phenylsulfanyl)-4-[1,2,3-tris(acetyloxy)propyl]-tetrahydropyrano[3,4-d][1,3]oxazole-6-carboxylate
Molecular Weight (g/mol): 567.56
Molecular Formula: C25H29NO12S
InChi Key: CVMYPHWPKCZUMN-UHFFFAOYNA-N
SMILES: COC(=O)C1(CC2OC(=O)N(C2C(O1)C(OC(C)=O)C(COC(C)=O)OC(C)=O)C(C)=O)SC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results