CAS RN 931-49-7
IUPAC Name:
1-ethynylcyclohex-1-ene
Synonyms:
1-ethynylcyclohex-1-ene
Molecular Weight (g/mol):
106.17
Molecular Formula:
C8H10
InChi Key:
DKFHWNGVMWFBJE-UHFFFAOYSA-N
SMILES:
C#CC1=CCCCC1
1
–
2
of
2
results
1
1-Ethynyl-1-cyclohexene 98.0+%, TCI America™
CAS: 931-49-7 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00001568 InChI Key: DKFHWNGVMWFBJE-UHFFFAOYSA-N Synonym: (1-Cyclohexenyl)acetylene PubChem CID: 79128 IUPAC Name: 1-ethynylcyclohexene SMILES: C#CC1=CCCCC1
2
Sigma Aldrich 1-Ethynylcyclohexene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.