missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 1-Ethynylcyclohexene
Description
1-Ethynylcyclohexene
Specifications
Specifications
| CAS | 931-49-7 |
| Density | 0.903 g/mL at 25°C (lit.) |
| Boiling Point | 148°C to 151°C (lit.) |
| Molecular Formula | C8H10 |
| Linear Formula | C6H9C///CH |
| Refractive Index | n20/D 1.496 (lit.) |
| Quantity | 25 g |
| MDL Number | MFCD00001568 |
| Molecular Weight (g/mol) | 106.17 |
| Percent Purity | 99% |
| Show More |