missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-1-cyclohexene 98.0+%, TCI America™

Catalog No. E11785ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11785ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11785ML Supplier TCI America Supplier No. E11785ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 931-49-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00001568
InChI Key DKFHWNGVMWFBJE-UHFFFAOYSA-N
Synonym (1-Cyclohexenyl)acetylene
PubChem CID 79128
IUPAC Name 1-ethynylcyclohexene
SMILES C#CC1=CCCCC1

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 mL
UN Number 3295
Formula Weight 106.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-1-cyclohexene
TSCA No
Recommended Storage Refrigerator