accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 931-19-1

IUPAC Name: 2-methylpyridin-1-ium-1-olate
Synonyms: picoline N-oxide methylpyridine 1-oxide
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: CFZKDDTWZYUZKS-UHFFFAOYSA-N
SMILES: CC1=CC=CC=[N+]1[O-]
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)