missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine N-Oxide 99.0+%, TCI America™

Catalog No. P0418500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0418500G 500 g
P041825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0418500G Supplier TCI America Supplier No. P0418500G

Chemical Identifiers

CAS 931-19-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006199
InChI Key CFZKDDTWZYUZKS-UHFFFAOYSA-N
Synonym 2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide
PubChem CID 13602
ChEBI CHEBI:35578
IUPAC Name 2-methyl-1-oxidopyridin-1-ium
SMILES CC1=CC=CC=[N+]1[O-]

Specifications

Color White-Yellow
Boiling Point 261°C
Quantity 500 g
Formula Weight 109.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methylpyridine N-Oxide