missing translation for 'onlineSavingsMsg'
Learn More

2-Picoline N-oxide, 98%

Catalog No. AAL0448836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0448836 500 g
AAL0448814 25 g
AAL0448822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0448836 Supplier Thermo Scientific Chemicals Supplier No. L0448836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 931-19-1 | C6H7NO | 109.128 g/mol

2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates.

Chemical Identifiers

CAS 931-19-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006199
InChI Key CFZKDDTWZYUZKS-UHFFFAOYSA-N
Synonym 2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide
PubChem CID 13602
ChEBI CHEBI:35578
IUPAC Name 2-methyl-1-oxidopyridin-1-ium
SMILES CC1=CC=CC=[N+]1[O-]

Specifications

Melting Point 46°C to 50°C
Boiling Point 122°C (8 mmHg)
Flash Point 143°C (289°F)
Quantity 500 g
Beilstein 106916
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Picoline N-oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-230-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only