accessibility menu, dialog, popup

UserName

CAS RN 930111-02-7

CH3NNNOOH

CAS RN 930111-02-7

IUPAC Name: 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylic acid
Synonyms: 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylic acid
Molecular Weight (g/mol): 221.26
Molecular Formula: C11H15N3O2
InChi Key: TTWSTFTZNIMXTA-UHFFFAOYNA-N
SMILES: CC1=CN=CC(=N1)N1CCCC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)