missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Methylpyrazin-2-yl)piperidine-3-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 930111-02-7
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD09879914
InChI Key TTWSTFTZNIMXTA-UHFFFAOYSA-N
Synonym 1-6-methylpyrazin-2-yl piperidine-3-carboxylic acid
PubChem CID 24229596
IUPAC Name 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylic acid
SMILES CC1=CN=CC(=N1)N2CCCC(C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC59001CB
SDS View Documents
Thermo Scientific Chemicals
CC59001CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59001DA
SDS View Documents
Thermo Scientific Chemicals
CC59001DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

930111-02-7
221.26
TTWSTFTZNIMXTA-UHFFFAOYSA-N
24229596
CC1=CN=CC(=N1)N2CCCC(C2)C(=O)O
C11H15N3O2
MFCD09879914
1-6-methylpyrazin-2-yl piperidine-3-carboxylic acid
1-(6-methylpyrazin-2-yl)piperidine-3-carboxylic acid

Specifications

930111-02-7
100.0
Amber Glass Bottle
MFCD09879914
1-6-methylpyrazin-2-yl piperidine-3-carboxylic acid
CC1=CN=CC(=N1)N2CCCC(C2)C(=O)O
221.26
221.26
1-(6-Methylpyrazin-2-yl)piperidine-3-carboxylic acid
96.5
97%
C11H15N3O2
250mg
TTWSTFTZNIMXTA-UHFFFAOYSA-N
1-(6-methylpyrazin-2-yl)piperidine-3-carboxylic acid
24229596
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents