accessibility menu, dialog, popup

UserName

CAS RN 93-04-9

IUPAC Name: 2-methoxynaphthalene
Synonyms: methyl β-naphthyl ether Β-naphthol methyl ether
Molecular Weight (g/mol): 158.2
Molecular Formula: C11H10O
InChi Key: LUZDYPLAQQGJEA-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)