missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynaphthalene, 98%, Thermo Scientific™

Catalog No. p-157556
Change view
Click to view available options
Quantity:
5g
500g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164750050 5g Glass bottle
AC164755000 500g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164750050 Supplier Thermo Scientific Chemicals Supplier No. 164750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 93-04-9
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004061
InChI Key LUZDYPLAQQGJEA-UHFFFAOYSA-N
Synonym naphthalene, 2-methoxy, yara yara, nerolin, methyl 2-naphthyl ether, yura yara, 2-naphthyl methyl ether, yara-yara, 2-naphthol methyl ether, nerolin old, beta-naphthyl methyl ether
PubChem CID 7119
IUPAC Name 2-methoxynaphthalene
SMILES COC1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 274°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C10H7OCH3
Quantity 5g
Beilstein 06, 640
Merck Index 15, 6070
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,carbon disulfide and,benzene
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material 2-Methoxynaphthalene, 98%
Show More Show Less
EINECSNumber 202-213-6
RTECSNumber QJ9468750
TSCA TSCA

RUO – Research Use Only