accessibility menu, dialog, popup

UserName

H2NH2N

CAS RN 92821-64-2

IUPAC Name: pyrene-1,3-diamine
Synonyms: pyrene-1,3-diamine
Molecular Weight (g/mol): 232.29
Molecular Formula: C16H12N2
InChi Key: WOFKFNZIJZWWPZ-UHFFFAOYSA-N
SMILES: NC1=CC(N)=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results