missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hexen-1-ol 95.0+%, TCI America™

Catalog No. H075410ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H075410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H075410ML Supplier TCI America Supplier No. H075410ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 928-91-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00148974,MFCD00009713
InChI Key VTIODUHBZHNXFP-IHWYPQMZSA-N
Synonym cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol #
PubChem CID 5365589
IUPAC Name (4Z)-hex-4-en-1-ol
SMILES C\C=C/CCCO

Specifications

Color Yellow
Boiling Point 158°C
Quantity 10 mL
Formula Weight 100.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-4-Hexen-1-ol
EINECSNumber (2)-2393
TSCA Yes