accessibility menu, dialog, popup

UserName

OHHC

CAS RN 927-74-2

IUPAC Name: but-3-yn-1-ol
Synonyms: 3-butyn-1-ol homopropargyl alcohol
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: OTJZCIYGRUNXTP-UHFFFAOYSA-N
SMILES: OCCC#C
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)