accessibility menu, dialog, popup

UserName

CAS RN 92-52-4

IUPAC Name: 1,1'-biphenyl
Synonyms: diphenyl caswell no. 087 14c-labeled diphenyl diphenyl, 14C-labeled xenene dowtherm bibenzene epa pesticide chemical code 017002 tetrosin LY phenador-X
Molecular Weight (g/mol): 154.21
Molecular Formula: C12H10
InChi Key: ZUOUZKKEUPVFJK-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)