accessibility menu, dialog, popup

UserName

FFFFFFFNNNFFFFFFFFFFFFFF

CAS RN 915-76-4

IUPAC Name: tris(1,1,2,2,3,3,3-heptafluoropropyl)-1,3,5-triazine
Synonyms: tris(1,1,2,2,3,3,3-heptafluoropropyl)-1,3,5-triazine
Molecular Weight (g/mol): 585.12
Molecular Formula: C12F21N3
InChi Key: KXQUYHRRCVECPV-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C1=NC(=NC(=N1)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)