missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.

Catalog No. AAL1688306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1688306 5 g
AAL1688303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1688306 Supplier Thermo Scientific Chemicals Supplier No. L1688306

missing translation for 'validMainframeMsg'

CAS: 915-76-4 | C12F21N3 | 585.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 915-76-4
Molecular Formula C12F21N3
Molecular Weight (g/mol) 585.119
MDL Number MFCD00042438
InChI Key KXQUYHRRCVECPV-UHFFFAOYSA-N
Synonym tris heptafluoropropyl-s-triazine, s-triazine, 2,4,6-tris heptafluoropropyl, 2,4,6-tris heptafluoropropyl-1,3,5-triazine, tris perfluoropropyl-s-triazine, 1,3,5-triazine, 2,4,6-tris heptafluoropropyl, 2,4,6-tris perfluoropropyl-1,3,5-triazine, perfluorotripropyl-s-triazine, tris heptafluoropropyl-1,3,5-triazine, s-triazine,4,6-tris heptafluoropropyl, 1,5-triazine, 2,4,6-tris heptafluoropropyl
PubChem CID 70185
IUPAC Name 2,4,6-tris(1,1,2,2,3,3,3-heptafluoropropyl)-1,3,5-triazine
SMILES C1(=NC(=NC(=N1)C(C(C(F)(F)F)(F)F)(F)F)C(C(C(F)(F)F)(F)F)(F)F)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.716
Boiling Point 165°C
Refractive Index 1.311
Quantity 5 g
Beilstein 381010
Formula Weight 585.12
Chemical Name or Material 2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Standard
EINECSNumber 213-023-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only