missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noscapine hydrochloride hydrate, MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| Chemical Name or Material | Noscapine Hydrochloride |
| CAS | 912-60-7 |
| Physical Form | Neat |
| Linear Formula | C22H23 NO7 · HCl · xH2 O |
| Technique | Liquid Chromatography |
| MDL Number | MFCD00149223 |
| Synonym | Narcotine hydrochloride |
| Type | Analytical Standard |
| Molecular Weight (g/mol) | 449.88 |
| Industry Type | Drug Analysis |
| Show More |