accessibility menu, dialog, popup

UserName

CH3OHNOOPOOHOPOOHOONOHNOOHOHHOOHOHNaNa

CAS RN 91183-98-1

IUPAC Name: {[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}[({[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}(hydroxy)phosphoryl)oxy]phosphinic acid disodium
Synonyms: [5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy({[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy(hydroxy)phosphoryl}oxy)phosphinic acid disodium
Molecular Weight (g/mol): 653.33
Molecular Formula: C17H27N3Na2O17P2
InChi Key: QPWKUQXSMYTKRZ-UHFFFAOYNA-N
SMILES: [Na].[Na].CC(=O)NC1C(O)C(O)C(CO)OC1OP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results