missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ UDP-α-D-N-Acetylglucosamine, Disodium Salt, >95% Calbiochem™,
Specifications
Specifications
| Color | White |
| Quantity | 50 mg |
| Solubility Information | Easily soluble in cold water |
| Formula Weight | 651.3 |
| Grade | HPLC |
| Physical Form | Solid |
| Chemical Name or Material | UDP-α-D-N-Acetylglucosamine, Disodium Salt |
Safety and Handling
| Recommended Storage | Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F). |