accessibility menu, dialog, popup

UserName

CAS RN 91170-93-3

IUPAC Name: 3-chloro-4-fluorobenzene-1-sulfonyl chloride
Synonyms: 3-chloro-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 229.05
Molecular Formula: C6H3Cl2FO2S
InChi Key: DXFXNSNBZNELII-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Cl)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)