missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzenesulfonyl chloride, 98%

Catalog No. AAA1122214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1122214 25 g
AAA1122206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1122214 Supplier Thermo Scientific Chemicals Supplier No. A1122214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91170-93-3 | C6H3Cl2FO2S | 229.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91170-93-3
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD00041444
InChI Key DXFXNSNBZNELII-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzene-1-sulfonyl chloride, 3-chloro-4-fluorobenzenesulphonyl chloride, 3-chloro-4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro-4-fluoro, 3-chloro-4-fluorophenylsulfonyl chloride, 3-chloro-4-fluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-chloro-4-fluoro-9ci, pubchem5093, acmc-209r8d, ksc488g2p
PubChem CID 145222
IUPAC Name 3-chloro-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)Cl)F

Specifications

Density 1.61
Boiling Point 104°C to 105°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.552
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only