missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. C17725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17725G 5 g
C177225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17725G Supplier TCI America Supplier No. C17725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91170-93-3
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD00041444
InChI Key DXFXNSNBZNELII-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzene-1-sulfonyl chloride, 3-chloro-4-fluorobenzenesulphonyl chloride, 3-chloro-4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro-4-fluoro, 3-chloro-4-fluorophenylsulfonyl chloride, 3-chloro-4-fluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-chloro-4-fluoro-9ci, pubchem5093, acmc-209r8d, ksc488g2p
PubChem CID 145222
IUPAC Name 3-chloro-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)Cl)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 229.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorobenzenesulfonyl Chloride