accessibility menu, dialog, popup

UserName

H3CHN

CAS RN 91-61-2

IUPAC Name: 6-methyl-1,2,3,4-tetrahydroquinoline
Synonyms: tetrahydro-6-methylquinoline
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: XOKMRXSMOHCNIX-UHFFFAOYSA-N
SMILES: CC1=CC=C2NCCCC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)