accessibility menu, dialog, popup

UserName

CAS RN 91-57-6

IUPAC Name: 2-methylnaphthalene
Synonyms: 2-methylnaphthalene 2-methylnaphthalene, ion(1-) 2-methylnaphthalene, ion(1+) ion(1-) 2-methylnaphthalene ion(1+) 2-methylnaphthalene
Molecular Weight (g/mol): 142.2
Molecular Formula: C11H10
InChi Key: QIMMUPPBPVKWKM-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)