accessibility menu, dialog, popup

UserName

ONH(R)O

CAS RN 90319-52-1

IUPAC Name: (4R)-4-phenyl-1,3-oxazolidin-2-one
Synonyms: (4R)-4-phenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: QDMNNMIOWVJVLY-QMMMGPOBSA-N
SMILES: O=C1N[C@@H](CO1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)