accessibility menu, dialog, popup

UserName

CAS RN 901245-65-6

H3CNNHOOH3CNNOH3C

CAS RN 901245-65-6

IUPAC Name: N-[1,3-dimethyl-2-oxo-6-(piperidin-1-yl)-2,3-dihydro-1H-1,3-benzodiazol-5-yl]-2-methoxybenzamide
Synonyms: N-[1,3-dimethyl-2-oxo-6-(piperidin-1-yl)-1,3-benzodiazol-5-yl]-2-methoxybenzamide
Molecular Weight (g/mol): 394.48
Molecular Formula: C22H26N4O3
InChi Key: LTUGYAOMCKNTGG-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NC1=C(C=C2N(C)C(=O)N(C)C2=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results