accessibility menu, dialog, popup

UserName

H3CONHOHOOSOHOOOOOOOHOOOHHOOSOHOOOHOOSHOOOHNSHOOOHOOSHOOOOHOHO

CAS RN 9005-49-6

IUPAC Name: 3-[(5-{[6-carboxy-4,5-dihydroxy-3-(sulfooxy)oxan-2-yl]oxy}-6-(hydroxymethyl)-3-(sulfoamino)-4-(sulfooxy)oxan-2-yl)oxy]-6-({5-acetamido-4,6-dihydroxy-2-[(sulfooxy)methyl]oxan-3-yl}oxy)-4-hydroxy-5-(sulfooxy)oxane-2-carboxylic acid
Synonyms: heparin enoxaparin sodium tinzaparin sodium bemiparin sodium parnaparin sodium
Molecular Weight (g/mol): 1134.9
Molecular Formula: C26H42N2O37S5
InChi Key: HTTJABKRGRZYRN-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)OC(COS(O)(=O)=O)C(OC2OC(C(OC3OC(CO)C(OC4OC(C(O)C(O)C4OS(O)(=O)=O)C(O)=O)C(OS(O)(=O)=O)C3NS(O)(=O)=O)C(O)C2OS(O)(=O)=O)C(O)=O)C1O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results