missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Heparin, Sodium Salt, Porcine Intestinal Mucosa, Calbiochem™,
Specifications
Specifications
| Packaging | Glass Bottle |
| Quantity | 500 kU |
| Grade | Not Available |
| Physical Form | Lyophilized |