accessibility menu, dialog, popup

UserName

NFFFFF

CAS RN 89402-42-6

IUPAC Name: 2,3-difluoro-5-(trifluoromethyl)pyridine
Synonyms: 2,3-difluoro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 183.08
Molecular Formula: C6H2F5N
InChi Key: XIFCGIKPAAZFFS-UHFFFAOYSA-N
SMILES: FC1=C(F)N=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)