missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-5-(trifluoromethyl)pyridine, 98%

Catalog No. AAH2595814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2595814 25 g
AAH2595803 1 g
AAH2595806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2595814 Supplier Thermo Scientific Chemicals Supplier No. H2595814
Only null left

CAS: 89402-42-6 | C6H2F5N | 183.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89402-42-6
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD04972837
InChI Key XIFCGIKPAAZFFS-UHFFFAOYSA-N
Synonym 2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine
PubChem CID 13421547
IUPAC Name 2,3-difluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(F)N=CC(=C1)C(F)(F)F

Specifications

Density 1.47
Boiling Point 104°C
Flash Point >100°C (212°F)
Quantity 25 g
Formula Weight 183.08
Percent Purity 98%
Chemical Name or Material 2,3-Difluoro-5-(trifluoromethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only