accessibility menu, dialog, popup

UserName

CH3ONNNHOOOCH3

CAS RN 89392-03-0

IUPAC Name: phenyl N-(4,6-dimethoxypyrimidin-2-yl)carbamate
Synonyms: phenyl N-(4,6-dimethoxypyrimidin-2-yl)carbamate
Molecular Weight (g/mol): 275.26
Molecular Formula: C13H13N3O4
InChi Key: MESPVSMSORHLAX-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(NC(=O)OC2=CC=CC=C2)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results