accessibility menu, dialog, popup

UserName

CAS RN 89-65-6

IUPAC Name: sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
Synonyms: sodium ascorbate
Molecular Weight (g/mol): 198.11
Molecular Formula: C6H7NaO6
InChi Key: IFVCRSPJFHGFCG-HXPAKLQESA-N
SMILES: [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)