missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Isoascorbic acid, 98%

Catalog No. AC227275000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227275000 500 g
AC227270025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC227275000 Supplier Thermo Scientific Chemicals Supplier No. 227275000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00005378
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
PubChem CID 54675810
ChEBI CHEBI:51438
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 167.0°C to 172.0°C
Color White to Yellow
Loss on Drying 0.4% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 5171
Specific Rotation Condition −17.25°(25°C c=10,H2O)
Specific Rotation − 17.25
Solubility Information Solubility in water: 100g/l. Other solubilities: soluble in alcohol,pyridine,moderately soluble in acetone,slightly soluble in glycerol
Formula Weight 176.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Isoascorbic acid, 98%
Show More Show Less
EINECSNumber 201-928-0
RTECSNumber KF3015000
TSCA TSCA

RUO – Research Use Only