accessibility menu, dialog, popup

UserName

CAS RN 88938-12-9

H3COHH3CHO

CAS RN 88938-12-9

IUPAC Name: 4-[9-(4-hydroxy-3-methylphenyl)-9H-fluoren-9-yl]-2-methylphenol
Synonyms: 4-[9-(4-hydroxy-3-methylphenyl)fluoren-9-yl]-2-methylphenol
Molecular Weight (g/mol): 378.47
Molecular Formula: C27H22O2
InChi Key: NUDSREQIJYWLRA-UHFFFAOYSA-N
SMILES: CC1=C(O)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(C)=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results