CAS RN 882-26-8
IUPAC Name:
1-nitro-3-[(1E)-2-nitroethenyl]benzene
Synonyms:
3-nitro-B-nitrostyrene
Molecular Weight (g/mol):
194.15
Molecular Formula:
C8H6N2O4
InChi Key:
YOEGXQQUPVDQEE-SNAWJCMRSA-N
SMILES:
[O-][N+](=O)\C=C\C1=CC=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
beta,3-Dinitrostyrene, 98%
beta,3-Dinitrostyrene, 98%
CAS: 882-26-8 Molecular Formula: C8H6N2O4 Molecular Weight (g/mol): 194.146 MDL Number: MFCD00052063 InChI Key: YOEGXQQUPVDQEE-SNAWJCMRSA-N Synonym: beta,3-dinitrostyrene,1-nitro-3-2-nitrovinyl benzene,m,.beta.-dinitrostyrene,trans-m,beta-dinitrostyrene,m-2-nitrovinyl nitrobenzene,3-nitro-b-nitrostyrene,styrene, m,beta-dinitro-, e,benzene, 1-nitro-3-2-nitroethenyl,1-nitro-2-m-nitrophenyl ethylene,1-nitro-3-e-2-nitroethenyl benzene PubChem CID: 5355017 IUPAC Name: 1-nitro-3-[(E)-2-nitroethenyl]benzene SMILES: C1=CC(=CC(=C1)[N+](=O)[O-])C=C[N+](=O)[O-]