accessibility menu, dialog, popup

UserName

CAS RN 88-42-6

OHSOOClCl

CAS RN 88-42-6

IUPAC Name: 2,5-dichlorobenzene-1-sulfonic acid
Synonyms: 2,5-dichlorobenzenesulfonic acid
Molecular Weight (g/mol): 227.06
Molecular Formula: C6H4Cl2O3S
InChi Key: LFXZSGVZSSMCMB-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results