missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzenesulfonic Acid Dihydrate 98.0+%, TCI America™

Catalog No. D033425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D033425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D033425G Supplier TCI America Supplier No. D033425G
Only null left

Chemical Identifiers

CAS 88-42-6
Molecular Formula C6H4Cl2O3S
Molecular Weight (g/mol) 227.055
MDL Number MFCD00035751
InChI Key LFXZSGVZSSMCMB-UHFFFAOYSA-N
PubChem CID 66616
IUPAC Name 2,5-dichlorobenzenesulfonic acid
SMILES C1=CC(=C(C=C1Cl)S(=O)(=O)O)Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 227.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorobenzenesulfonic Acid Dihydrate