missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylbutyric Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 193°C |
| Quantity | 500 mL |
| UN Number | 2810 |
| Formula Weight | 116.16 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Ethylbutyric Acid |
Safety and Handling
| EINECSNumber | (2)-0608 |
| RTECSNumber | ET1400000 |
| TSCA | Yes |